Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144844.2 | NP_659093.2 | 7 | 574 | ||
GenBank mRNA | AK148002.1 | - | 7 | 564 | ||
AY046947.1 | - | 7 | 591 | |||
BC006915.1 | - | 7 | 571 | 3601148 | ||
BC049802.1 | - | 7 | 540 | 6332730 |