Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TXNDC5 SIRNA Assays ›
Gene Symbol:
Txndc5
Gene Name:
thioredoxin domain containing 5
Gene Aliases:
AL022641, ERp46, PC-TRP
Chromosome Location:
Chr.13: 38500266 - 38528824 on Build GRCm38
UniGene ID:
Mm.28622
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
173264
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Txndc5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289598.1
NP_001276527.1
2
357
NM_001289599.1
NP_001276528.1
2
324
NM_145367.4
NP_663342.3
2
381
GenBank mRNA
AK136046.1
-
2
269
AK142049.1
-
2
367
AK150781.1
-
2
365
AK169680.1
-
2
348
AK224911.1
-
1
34
AK224913.1
-
2
368
AK224922.1
-
2
326
AK224923.1
-
2
348
AK224924.1
-
2
356
AK224926.1
-
2
326
AK224927.1
-
2
377
AK224930.1
-
2
329
AK224933.1
-
2
318
AK224934.1
-
2
372
AK224936.1
-
2
371
AK224937.1
-
2
370
AK224938.1
-
2
365
AK224939.1
-
2
358
AK224943.1
-
2
329
AK224945.1
-
2
365
AK224946.1
-
2
368
AK224947.1
-
2
355
AK224948.1
-
2
329
AK224949.1
-
2
318
AK224950.1
-
2
318
AK224951.1
-
2
376
AK224952.1
-
2
344
AK224956.1
-
2
289
AK224957.1
-
2
334
AK224958.1
-
2
334
AK224960.1
-
2
376
AK224962.1
-
2
336
AK224963.1
-
2
377
AK224964.1
-
2
377
AK224965.1
-
2
353
AK224966.1
-
2
353
AK224967.1
-
2
344
AK224970.1
-
2
318
AK224971.1
-
2
378
AK224972.1
-
2
368
AK224973.1
-
2
318
AK224974.1
-
2
321
AK224976.1
-
2
351
AK224977.1
-
2
365
AK224978.1
-
2
365
AK224979.1
-
2
352
AK224981.1
-
2
322
AK224982.1
-
2
377
AK224983.1
-
2
365
AK224984.1
-
2
331
AK224985.1
-
2
335
AK224986.1
-
2
379
AK224988.1
-
2
355
AK224989.1
-
2
372
AK224990.1
-
2
367
AK224991.1
-
2
362
AK224992.1
-
2
362
AK224993.1
-
2
362
AK224994.1
-
2
370
AK224995.1
-
2
333
AK224996.1
-
2
304
AK224997.1
-
2
329
AK224998.1
-
2
327
AK224999.1
-
2
349
AK225004.1
-
2
370
AK225005.1
-
2
348
AK225007.1
-
2
370
AK225010.1
-
2
370
AK225011.1
-
2
370
AK225012.1
-
2
139
AK225013.1
-
2
139
AK225014.1
-
2
348
AK225015.1
-
2
139
AK225016.1
-
2
278
AK225017.1
-
2
326
AK225019.1
-
2
358
AK225020.1
-
2
309
AK225021.1
-
2
309
AK225022.1
-
2
157
AK225023.1
-
2
382
AY243534.1
-
2
344
AY548112.1
-
2
305
BC016252.1
-
2
348
4911455
BC024505.1
-
2
293
4983496
BC046789.1
-
2
307
5716897
BP765107.1
-
2
371
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
response to endoplasmic reticulum stress
apoptotic cell clearance
cell redox homeostasis
protein disulfide isomerase activity
isomerase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0