Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TXNDC5 SIRNA Assays ›
Gene Symbol:
Txndc5
Gene Name:
thioredoxin domain containing 5
Gene Aliases:
AL022641, ERp46, PC-TRP
Chromosome Location:
Chr.13: 38500266 - 38528824 on Build GRCm38
UniGene ID:
Mm.28622
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
173265
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Txndc5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289598.1
NP_001276527.1
4
597
NM_001289599.1
NP_001276528.1
4
564
NM_145367.4
NP_663342.3
4
621
GenBank mRNA
AK136046.1
-
4
509
AK142049.1
-
4
607
AK150781.1
-
4
605
AK169680.1
-
4
588
AK224911.1
-
3
274
AK224913.1
-
4
608
AK224922.1
-
4
566
AK224923.1
-
4
588
AK224924.1
-
4
596
AK224926.1
-
4
566
AK224927.1
-
4
617
AK224930.1
-
4
569
AK224933.1
-
4
558
AK224934.1
-
4
612
AK224936.1
-
4
611
AK224937.1
-
4
610
AK224938.1
-
4
605
AK224939.1
-
4
598
AK224943.1
-
4
569
AK224945.1
-
4
605
AK224946.1
-
4
608
AK224947.1
-
4
595
AK224948.1
-
4
569
AK224949.1
-
4
558
AK224950.1
-
4
558
AK224951.1
-
4
616
AK224952.1
-
4
584
AK224956.1
-
4
529
AK224957.1
-
4
574
AK224958.1
-
4
574
AK224960.1
-
4
616
AK224962.1
-
4
576
AK224963.1
-
4
617
AK224964.1
-
4
617
AK224965.1
-
4
593
AK224966.1
-
4
593
AK224967.1
-
4
584
AK224968.1
-
2
151
AK224969.1
-
2
151
AK224970.1
-
4
558
AK224971.1
-
4
618
AK224972.1
-
4
608
AK224973.1
-
4
558
AK224974.1
-
4
561
AK224976.1
-
4
591
AK224977.1
-
4
605
AK224978.1
-
4
605
AK224979.1
-
4
592
AK224981.1
-
4
562
AK224982.1
-
4
617
AK224983.1
-
4
606
AK224984.1
-
4
571
AK224985.1
-
4
575
AK224986.1
-
4
619
AK224988.1
-
4
595
AK224989.1
-
4
612
AK224990.1
-
4
607
AK224991.1
-
4
602
AK224992.1
-
4
602
AK224993.1
-
4
602
AK224994.1
-
4
610
AK224995.1
-
4
573
AK224996.1
-
4
544
AK224997.1
-
4
569
AK224998.1
-
4
567
AK224999.1
-
4
589
AK225004.1
-
4
610
AK225005.1
-
4
588
AK225007.1
-
4
610
AK225010.1
-
4
610
AK225011.1
-
4
610
AK225012.1
-
4
379
AK225013.1
-
4
379
AK225014.1
-
4
588
AK225015.1
-
4
379
AK225016.1
-
4
518
AK225017.1
-
4
566
AK225019.1
-
4
598
AK225020.1
-
4
549
AK225021.1
-
4
549
AK225022.1
-
4
397
AK225023.1
-
4
623
AY243534.1
-
4
584
AY548112.1
-
4
545
BC016252.1
-
4
588
4911455
BC024505.1
-
4
533
4983496
BC036155.1
-
3
207
BC046789.1
-
4
547
5716897
BP765107.1
-
4
611
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001271330, FM067602.
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
response to endoplasmic reticulum stress
apoptotic cell clearance
cell redox homeostasis
protein disulfide isomerase activity
isomerase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0