Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Txndc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289598.1 | NP_001276527.1 | 10 | 2108 | ||
NM_001289599.1 | NP_001276528.1 | 10 | 2075 | |||
NM_145367.4 | NP_663342.3 | 10 | 2132 | |||
GenBank mRNA | AK142049.1 | - | 10 | 2118 | ||
AK150781.1 | - | 10 | 2116 | |||
AK169680.1 | - | 10 | 2099 | |||
AK201879.1 | - | 1 | 199 | |||
AK224911.1 | - | 9 | 1785 | |||
AK224912.1 | - | 1 | 583 | |||
AK224913.1 | - | 10 | 2119 | |||
AK224914.1 | - | 1 | 515 | |||
AK224915.1 | - | 1 | 515 | |||
AK224916.1 | - | 1 | 515 | |||
AK224917.1 | - | 1 | 515 | |||
AK224919.1 | - | 1 | 515 | |||
AK224920.1 | - | 1 | 515 | |||
AK224921.1 | - | 1 | 515 | |||
AK224922.1 | - | 10 | 2077 | |||
AK224923.1 | - | 10 | 2099 | |||
AK224924.1 | - | 10 | 2107 | |||
AK224926.1 | - | 10 | 2077 | |||
AK224927.1 | - | 10 | 2128 | |||
AK224930.1 | - | 10 | 2080 | |||
AK224933.1 | - | 10 | 2069 | |||
AK224934.1 | - | 10 | 2123 | |||
AK224935.1 | - | 3 | 2274 | |||
AK224936.1 | - | 10 | 2122 | |||
AK224937.1 | - | 10 | 2121 | |||
AK224938.1 | - | 10 | 2116 | |||
AK224939.1 | - | 10 | 2109 | |||
AK224943.1 | - | 10 | 2080 | |||
AK224945.1 | - | 10 | 2116 | |||
AK224946.1 | - | 10 | 2119 | |||
AK224947.1 | - | 10 | 2106 | |||
AK224948.1 | - | 10 | 2080 | |||
AK224949.1 | - | 10 | 2069 | |||
AK224950.1 | - | 10 | 2069 | |||
AK224951.1 | - | 10 | 2127 | |||
AK224953.1 | - | 1 | 725 | |||
AK224955.1 | - | 5 | 1380 | |||
AK224956.1 | - | 10 | 2039 | |||
AK224957.1 | - | 10 | 2085 | |||
AK224958.1 | - | 10 | 2085 | |||
AK224960.1 | - | 10 | 2127 | |||
AK224962.1 | - | 10 | 2087 | |||
AK224963.1 | - | 7 | 1047 | |||
AK224964.1 | - | 7 | 1047 | |||
AK224965.1 | - | 10 | 2104 | |||
AK224966.1 | - | 10 | 2104 | |||
AK224967.1 | - | 10 | 2095 | |||
AK224968.1 | - | 8 | 1662 | |||
AK224969.1 | - | 8 | 1662 | |||
AK224970.1 | - | 10 | 2069 | |||
AK224971.1 | - | 10 | 2129 | |||
AK224972.1 | - | 10 | 2119 | |||
AK224973.1 | - | 10 | 2069 | |||
AK224974.1 | - | 10 | 2072 | |||
AK224975.1 | - | 5 | 1307 | |||
AK224976.1 | - | 10 | 2102 | |||
AK224977.1 | - | 10 | 2116 | |||
AK224978.1 | - | 10 | 2116 | |||
AK224979.1 | - | 10 | 2103 | |||
AK224981.1 | - | 10 | 2073 | |||
AK224982.1 | - | 10 | 2128 | |||
AK224983.1 | - | 10 | 2117 | |||
AK224984.1 | - | 10 | 2082 | |||
AK224985.1 | - | 10 | 2086 | |||
AK224986.1 | - | 10 | 2129 | |||
AK224988.1 | - | 10 | 2106 | |||
AK224989.1 | - | 10 | 2123 | |||
AK224990.1 | - | 10 | 2118 | |||
AK224991.1 | - | 10 | 2113 | |||
AK224992.1 | - | 10 | 2113 | |||
AK224993.1 | - | 10 | 2113 | |||
AK224994.1 | - | 10 | 2121 | |||
AK224995.1 | - | 10 | 2084 | |||
AK224997.1 | - | 10 | 2080 | |||
AK224998.1 | - | 10 | 2078 | |||
AK224999.1 | - | 10 | 2099 | |||
AK225000.1 | - | 1 | 893 | |||
AK225004.1 | - | 10 | 2121 | |||
AK225006.1 | - | 1 | 576 | |||
AK225007.1 | - | 10 | 2121 | |||
AK225008.1 | - | 1 | 576 | |||
AK225010.1 | - | 10 | 2121 | |||
AK225011.1 | - | 10 | 2121 | |||
AK225012.1 | - | 10 | 1890 | |||
AK225013.1 | - | 10 | 1890 | |||
AK225014.1 | - | 10 | 2099 | |||
AK225015.1 | - | 10 | 1890 | |||
AK225016.1 | - | 10 | 2029 | |||
AK225017.1 | - | 10 | 2077 | |||
AK225019.1 | - | 10 | 2109 | |||
AK225020.1 | - | 10 | 2060 | |||
AK225021.1 | - | 10 | 2060 | |||
AK225022.1 | - | 10 | 1908 | |||
AK225023.1 | - | 10 | 2134 | |||
AY243534.1 | - | 10 | 2095 | |||
BC016252.1 | - | 10 | 2099 | 4911455 | ||
BC036155.1 | - | 9 | 1718 | |||
BC046789.1 | - | 10 | 2058 | 5716897 |