Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145929.2 | NP_666041.1 | 10 | 1253 | ||
GenBank mRNA | AK080881.1 | - | 10 | 876 | ||
AK155195.1 | - | 10 | 893 | |||
AK159771.1 | - | 10 | 876 | |||
AK170437.1 | - | 10 | 894 | |||
AK171534.1 | - | 10 | 910 | |||
AK185927.1 | - | 3 | 229 | |||
AK218460.1 | - | 3 | 229 | |||
BC026802.1 | - | 10 | 1253 | 4223300 | ||
BC054422.1 | - | 10 | 897 |