Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145939.2 | NP_666051.2 | 7, 8 | 1013 | ||
GenBank mRNA | AK090067.1 | - | 6, 7 | 914 | ||
AK149510.1 | - | 6, 7 | 913 | |||
BC031893.1 | - | 7, 8 | 1008 | 5371807 |