Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kbtbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145958.2 | NP_666070.2 | 4 | 1345 | ||
GenBank mRNA | AK028396.1 | - | 4 | 1291 | ||
AK129374.1 | - | 4 | 1255 | |||
BC022962.1 | - | 1 | 262 | 5368323 | ||
BC023724.1 | - | 1 | 134 |