Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145979.2 | NP_666091.1 | 3 | 299 | ||
GenBank mRNA | AK020564.1 | - | 2 | 720 | ||
AK040005.1 | - | 3 | 341 | |||
AK132004.1 | - | 3 | 312 | |||
AK145532.1 | - | 3 | 286 | |||
AK147695.1 | - | 3 | 482 | |||
AK220473.1 | - | 2 | 186 | |||
BC058578.1 | - | 3 | 299 | 6489649 |