Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146061.4 | NP_666173.4 | 7 | 860 | ||
GenBank mRNA | AF482997.1 | - | 7 | 855 | ||
AK046057.1 | - | 8 | 677 | |||
AK081231.1 | - | 7 | 856 |