Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam210a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153794.4 | NP_722489.1 | 4 | 1371 | ||
GenBank mRNA | AK034980.1 | - | 4 | 1253 | ||
AK044238.1 | - | 4 | 1260 | |||
AK049567.1 | - | 4 | 1259 | |||
AK077093.1 | - | 4 | 1372 | |||
AK079137.1 | - | 1 | 494 | |||
AK142190.1 | - | 4 | 1243 | |||
AK147662.1 | - | 4 | 1245 | |||
AK159319.1 | - | 4 | 1220 | |||
AK162396.1 | - | 4 | 1257 | |||
BC038695.1 | - | 4 | 1139 | 1548421 | ||
BC095936.1 | - | 4 | 1220 | 3968496 |