Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172378.2 | NP_759010.1 | 7 | 1375 | ||
GenBank mRNA | AB520670.1 | - | 6 | 981 | ||
AK030222.1 | - | 7 | 1375 | |||
BC026439.1 | - | 6 | 1018 | 4239636 |