Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163755.1 | NP_001157227.1 | 9 | 3567 | ||
NM_001163757.1 | NP_001157229.1 | 9 | 3564 | |||
NM_001163758.1 | NP_001157230.1 | 9 | 3548 | |||
NM_172446.3 | NP_766034.2 | 9 | 3620 | |||
GenBank mRNA | AK029916.1 | - | 9 | 3551 | ||
AK049035.1 | - | 9 | 3621 | |||
AK134840.1 | - | 9 | 3556 |