Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bzrap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172449.2 | NP_766037.2 | 32 | 7393 | ||
GenBank mRNA | AK051895.1 | - | 10 | 3291 | ||
AK122329.1 | - | 9 | 2474 | |||
BC055691.1 | - | 12 | 2144 | |||
BC066146.1 | - | 10 | 2353 | 30470084 | ||
BC083181.1 | - | 12 | 2193 |