Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172468.2 | NP_766056.1 | 9 | 1939 | ||
GenBank mRNA | AK029012.1 | - | 9 | 1741 | ||
AK076374.1 | - | 6 | 943 | |||
AK147437.1 | - | 9 | 1941 | |||
AK165619.1 | - | 9 | 1749 | |||
AK170774.1 | - | 9 | 1713 | |||
AK170999.1 | - | 9 | 1741 | |||
AK216338.1 | - | 1 | 76 | |||
BC099674.1 | - | 9 | 1735 | 4950374 |