Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173748.4 | NP_776109.1 | 2 | 782 | ||
GenBank mRNA | AK036694.1 | - | 2 | 487 | ||
AK046219.1 | - | 2 | 504 | |||
AK049739.1 | - | 2 | 494 | |||
AK050476.1 | - | 2 | 472 | |||
AK159391.1 | - | 2 | 514 | |||
AK159458.1 | - | 2 | 514 | |||
AK169797.1 | - | 2 | 518 | |||
AK170329.1 | - | 2 | 515 | |||
AK171224.1 | - | 2 | 497 | |||
AK198358.1 | - | 1 | 14 | |||
BC024322.1 | - | 2 | 500 | 4236345 | ||
BC037090.1 | - | 2 | 481 | 5134044 | ||
BC052089.1 | - | 2 | 343 | |||
BC057603.1 | - | 1 | 250 | |||
EU350933.1 | - | 2 | 473 |