Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173748.4 | NP_776109.1 | 6 | 1820 | ||
GenBank mRNA | AK030050.1 | - | 6 | 1546 | ||
AK036694.1 | - | 6 | 1525 | |||
AK049739.1 | - | 6 | 1532 | |||
AK050476.1 | - | 5 | 1366 | |||
AK129276.1 | - | 3 | 735 | |||
AK159391.1 | - | 6 | 1553 | |||
AK159458.1 | - | 6 | 1553 | |||
AK169797.1 | - | 6 | 1556 | |||
AK170329.1 | - | 6 | 1553 | |||
AK171224.1 | - | 6 | 1535 | |||
AK207279.1 | - | 1 | 364 | |||
AK207975.1 | - | 1 | 368 | |||
AK214587.1 | - | 1 | 345 | |||
AK218061.1 | - | 1 | 368 | |||
BC024322.1 | - | 6 | 1538 | 4236345 | ||
BC037090.1 | - | 6 | 1519 | 5134044 | ||
BC052089.1 | - | 6 | 1381 | |||
BC057603.1 | - | 5 | 1288 |