Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199360.1 | NP_001186289.1 | 2 | 763 | ||
NM_177592.4 | NP_808260.2 | 2 | 706 | |||
GenBank mRNA | AK081890.1 | - | 1 | 953 | ||
AK089342.1 | - | 2 | 613 | |||
AK165839.1 | - | 2 | 599 | |||
AK165910.1 | - | 2 | 599 | |||
BC056470.1 | - | 2 | 688 | 5706838 |