Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177594.1 | NP_808262.1 | 2 | 583 | ||
GenBank mRNA | AK035026.1 | - | 2 | 598 | ||
AK044957.1 | - | 2 | 613 | |||
AK152320.1 | - | 2 | 369 | |||
BC046275.1 | - | 2 | 583 | 6307469 | ||
BC145946.1 | - | 2 | 338 | 40131249 | ||
BC145948.1 | - | 2 | 338 | 40131251 |