Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177594.1 | NP_808262.1 | 10 | 1861 | ||
GenBank mRNA | AF073881.1 | - | 3 | 384 | ||
AK035026.1 | - | 10 | 1876 | |||
AK044957.1 | - | 10 | 1891 | |||
AK152320.1 | - | 10 | 1647 | |||
BC046275.1 | - | 10 | 1861 | 6307469 | ||
BC145946.1 | - | 10 | 1616 | 40131249 | ||
BC145948.1 | - | 10 | 1616 | 40131251 |