Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182997.2 | NP_892042.2 | 8 | 985 | ||
GenBank mRNA | AK037124.1 | - | 3 | 319 | ||
BC054401.1 | - | 2 | 152 | |||
BC060228.1 | - | 8 | 985 | 6839848 |