Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ccnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183015.3 | NP_898836.2 | 6, 7 | 752 | ||
GenBank mRNA | AJ555464.1 | - | 6, 7 | 753 | ||
BC132411.1 | - | 5, 6 | 706 | 40130688 | ||
BC138355.1 | - | 5, 6 | 706 | 8861375 |