Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C030039L03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112731.1 | NP_001106202.1 | 6 | 2134 | ||
NM_198417.2 | NP_940809.2 | 6 | 2114 | |||
GenBank mRNA | AK140966.1 | - | 6 | 2038 | ||
BC060708.1 | - | 6 | 2114 | 6406744 |
Gene Symbol: Zfp607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024726.2 | NP_001019897.2 | 5 | 2084 | ||
GenBank mRNA | AK028967.1 | - | 5 | 2063 | ||
AK143371.1 | - | 5 | 2052 | |||
AK163602.1 | - | 5 | 2084 | |||
BC042662.1 | - | 3740 | ||||
BC141161.1 | - | 5 | 2051 | 9055773 |