Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163759.1 | NP_001157231.1 | 5 | 1253 | ||
NM_198942.2 | NP_945180.2 | 4 | 1094 | |||
GenBank mRNA | AK053628.1 | - | 5 | 921 | ||
BC062952.1 | - | 4 | 1094 | 6853031 | ||
BC065169.1 | - | 5 | 920 | 6831314 | ||
BC066091.1 | - | 4 | 756 | 6841207 |