Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199447.2 | NP_955518.1 | 18 | 2234 | ||
GenBank mRNA | AK122346.1 | - | 17 | 3348 | ||
AK146550.1 | - | 18 | 2242 | |||
BC056232.1 | - | 18 | 2266 | 30060906 | ||
BC062977.1 | - | 18 | 2234 | 6853905 |