Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB028272.1 | - | 3 | 1932 | ||
AK146657.1 | - | 3 | 1881 | |||
AK158530.1 | - | 3 | 1778 | |||
AK167718.1 | - | 3 | 1818 | |||
AK168220.1 | - | 3 | 1832 | |||
AK168291.1 | - | 3 | 1832 | |||
AK185089.1 | - | 1 | 330 | |||
BC012962.1 | - | 3 | 1815 | 4188579 |