Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018855.2 | NP_061343.2 | 4 | 447 | ||
GenBank mRNA | AK140135.1 | - | 4 | 426 | ||
AY692443.1 | - | 4 | 446 | |||
BC003779.1 | - | 4 | 362 | 3592695 | ||
DQ189098.1 | - | 3 | 343 | |||
U19859.1 | - | 4 | 468 |