Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE4D SIRNA Assays ›
Gene Symbol:
PDE4D
Gene Name:
phosphodiesterase 4D
Gene Aliases:
ACRDYS2, DPDE3, HSPDE4D, PDE43, PDE4DN2, STRK1
Chromosome Location:
Chr.5: 58969038 - 60488098 on Build GRCh38
UniGene ID:
Hs.117545
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
17429
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE4D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001104631.1
NP_001098101.1
7
1109
NM_001165899.1
NP_001159371.1
9
1000
NM_001197218.1
NP_001184147.1
7
848
NM_001197219.1
NP_001184148.1
7
626
NM_001197220.1
NP_001184149.1
7
1264
NM_001197221.1
NP_001184150.1
3
584
NM_001197222.1
NP_001184151.1
2
670
NM_001197223.1
NP_001184152.1
2
460
NM_006203.4
NP_006194.2
7
652
XM_005248537.3
XP_005248594.1
7
1806
XM_005248538.4
XP_005248595.1
8
3707
XM_011543469.1
XP_011541771.1
10
1089
XM_011543470.2
XP_011541772.1
10
1013
XM_011543471.1
XP_011541773.1
9
966
XM_011543473.1
XP_011541775.1
9
914
XM_011543477.2
XP_011541779.1
9
916
XM_017009565.1
XP_016865054.1
10
1126
XM_017009566.1
XP_016865055.1
10
1016
XM_017009567.1
XP_016865056.1
8
754
XM_017009568.1
XP_016865057.1
10
1000
XM_017009569.1
XP_016865058.1
9
879
XM_017009570.1
XP_016865059.1
6
1699
XM_017009571.1
XP_016865060.1
9
922
GenBank mRNA
AF012073.1
-
7
862
AF012074.1
-
3
598
AF536975.1
-
2
267
AF536976.1
-
9
823
AF536977.1
-
7
626
AJ250852.1
-
3
194
AJ250854.1
-
8
789
AJ250855.1
-
6
487
AY245866.1
-
9
965
AY245867.1
-
7
1264
AY388960.1
-
7
547
BC036319.1
-
2
462
4828512
L20970.1
-
7
660
U02882.1
-
6
430
U50157.1
-
2
268
U50158.1
-
3
182
U50159.1
-
7
603
U79571.1
-
2
673
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00174810_m1
Panther Classification:
Molecular Function -
phosphodiesterase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart rate
cAMP catabolic process
smooth muscle contraction
aging
regulation of receptor activity
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
cAMP-mediated signaling
neutrophil chemotaxis
regulation of cAMP metabolic process
positive regulation of interferon-gamma production
positive regulation of interleukin-2 production
positive regulation of interleukin-5 production
negative regulation of peptidyl-serine phosphorylation
multicellular organism growth
negative regulation of heart contraction
T cell receptor signaling pathway
regulation of ryanodine-sensitive calcium-release channel activity
establishment of endothelial barrier
cellular response to lipopolysaccharide
cellular response to epinephrine stimulus
adrenergic receptor signaling pathway
regulation of cardiac muscle cell contraction
adrenergic receptor signaling pathway involved in positive regulation of heart rate
regulation of cell communication by electrical coupling involved in cardiac conduction
negative regulation of relaxation of cardiac muscle
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
protein binding
drug binding
enzyme binding
cAMP binding
beta-2 adrenergic receptor binding
ion channel binding
metal ion binding
ATPase binding
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0