Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199227.1 | NP_001186156.1 | 14 | 1701 | ||
NM_024289.2 | NP_077251.1 | 14 | 1901 | |||
GenBank mRNA | AB074008.1 | - | 14 | 1694 | ||
AJ278263.2 | - | 14 | 1901 | |||
AK129385.1 | - | 5 | 657 | |||
BC079872.1 | - | 14 | 1682 | 6832073 |