Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024467.3 | NP_077787.3 | 2 | 3611 | ||
GenBank mRNA | AK048624.1 | - | 2 | 3472 | ||
AK052274.1 | - | 2 | 3446 | |||
AK089263.1 | - | 2 | 3462 | |||
AK122500.1 | - | 2773 | ||||
BC033441.1 | - | 2 | 3440 | |||
BC059823.1 | - | 2 | 3611 | 6843792 |