Gene Symbol: Dtnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025772.4 | NP_080048.2 | 3 | 253 | ||
GenBank mRNA | AJ404859.1 | - | 3 | 252 | ||
AK150306.1 | - | 3 | 249 | |||
AK159656.1 | - | 3 | 378 | |||
BC018350.1 | - | 3 | 244 | 4195234 | ||
BC048682.1 | - | 3 | 298 | |||
BC058574.1 | - | 3 | 226 | 6487720 |