Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025772.4 | NP_080048.2 | 7, 8 | 643 | ||
GenBank mRNA | AJ404859.1 | - | 7, 8 | 642 | ||
AK010924.1 | - | 3, 4 | 255 | |||
AK017400.1 | - | 3, 4 | 421 | |||
AK159656.1 | - | 7, 8 | 768 | |||
AK189615.1 | - | 3, 4 | 205 | |||
AY265461.1 | - | 3, 4 | 421 | |||
BC018350.1 | - | 7, 8 | 634 | 4195234 | ||
BC048682.1 | - | 7, 8 | 688 | |||
BC058574.1 | - | 7, 8 | 616 | 6487720 |