Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025772.4 | NP_080048.2 | 10 | 1182 | ||
GenBank mRNA | AJ404859.1 | - | 10 | 1181 | ||
AK010924.1 | - | 6 | 794 | |||
AK017400.1 | - | 6 | 960 | |||
AK159656.1 | - | 10 | 1307 | |||
AK212317.1 | - | 2 | 286 | |||
AY265461.1 | - | 6 | 960 | |||
BC018350.1 | - | 10 | 1173 | 4195234 | ||
BC048682.1 | - | 10 | 1227 | |||
BC058574.1 | - | 10 | 1155 | 6487720 |