Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam192a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028221.4 | NP_082497.2 | 5, 6 | 685 | ||
GenBank mRNA | AK010053.1 | - | 5, 6 | 650 | ||
AK033988.1 | - | 5, 6 | 616 | |||
AK050704.1 | - | 5, 6 | 620 | |||
AK151603.1 | - | 5, 6 | 639 | |||
AK213161.1 | - | 1, 2 | 38 | |||
BC016088.1 | - | 4, 5 | 425 | 4503452 | ||
BC023158.1 | - | 5, 6 | 524 | 4946201 | ||
BC025472.1 | - | 5, 6 | 661 | 5353148 | ||
BC027060.1 | - | 5, 6 | 647 | 5358022 | ||
BC027818.1 | - | 4, 5 | 453 | 5320967 | ||
BC040818.1 | - | 638 | 4194531 |