Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029649.2 | NP_083925.2 | 4 | 776 | ||
GenBank mRNA | AK012106.1 | - | 4 | 730 | ||
AK041154.1 | - | 4 | 735 | |||
AK049378.1 | - | 4 | 758 | |||
AK140260.1 | - | 4 | 777 | |||
BC011109.1 | - | 4 | 746 | 2654288 |