Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029746.3 | NP_084022.2 | 6 | 625 | ||
GenBank mRNA | AK089920.1 | - | 6 | 609 | ||
AK157348.1 | - | 6 | 587 | |||
AK169298.1 | - | 6 | 595 | |||
AK171726.1 | - | 6 | 544 | |||
BC011435.1 | - | 6 | 565 | 4010102 | ||
CO423795.1 | - | 3 | 182 |