Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039201.1 | NP_001034290.1 | 7 | 992 | ||
NM_029826.2 | NP_084102.1 | 7 | 953 | |||
GenBank mRNA | AK014208.1 | - | 7 | 947 | ||
AK137285.1 | - | 7 | 945 | |||
AK147794.1 | - | 974 | ||||
BC058177.1 | - | 7 | 927 | 5250739 |