Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168403.1 | NP_001161875.1 | 3 | 669 | ||
NM_001168404.1 | NP_001161876.1 | 3 | 206 | |||
NM_001168405.1 | NP_001161877.1 | 2 | 563 | |||
NM_029898.3 | NP_084174.2 | 2 | 563 | |||
GenBank mRNA | AK039553.1 | - | 2 | 564 | ||
AK045480.1 | - | 2 | 197 | |||
AK081197.1 | - | 3 | 207 | |||
AK138273.1 | - | 2 | 218 | |||
AK140417.1 | - | 3 | 532 | |||
BC152317.1 | - | 2 | 190 | 40088531 | ||
BC152536.1 | - | 2 | 190 | 40088530 |