Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030132.5 | NP_084408.1 | 1, 2 | 275 | ||
GenBank mRNA | AK021306.1 | - | 1, 2 | 225 | ||
AK042518.1 | - | 1, 2 | 232 | |||
AK134445.1 | - | 1, 2 | 207 | |||
AK188127.1 | - | 1, 2 | 43 | |||
BC046791.1 | - | 1, 2 | 220 | 5717427 | ||
BC056961.1 | - | 1, 2 | 220 | 5717427 | ||
CK334116.1 | - | 1, 2 | 287 |