Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030132.5 | NP_084408.1 | 3 | 2229 | ||
GenBank mRNA | AK042518.1 | - | 3 | 2185 | ||
AK134445.1 | - | 3 | 2159 | |||
BC046791.1 | - | 3 | 2174 | 5717427 | ||
BC056961.1 | - | 3 | 2174 | 5717427 |