Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030701.3 | NP_109626.1 | 1 | 560 | ||
GenBank mRNA | AJ300198.1 | - | 1 | 540 | ||
AK150795.1 | - | 1 | 542 | |||
AK169956.1 | - | 1 | 544 | |||
BC120715.1 | - | 1 | 530 | 40129638 | ||
BC125296.1 | - | 1 | 530 | 40129811 |