Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl1xr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030732.3 | NP_109657.2 | 16 | 1984 | ||
GenBank mRNA | AF268195.1 | - | 16 | 1765 | ||
AK029595.1 | - | 16 | 1783 | |||
AK033347.1 | - | 16 | 1854 | |||
AK036064.1 | - | 17 | 2095 | |||
AK136255.1 | - | 16 | 1806 | |||
BC018512.1 | - | 6 | 630 |