Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031258.3 | NP_112548.2 | 9 | 1609 | ||
GenBank mRNA | AF296451.1 | - | 9 | 1531 | ||
AF305714.1 | - | 8 | 1148 | |||
AK142805.1 | - | 9 | 1233 | |||
AK164661.1 | - | 9 | 1204 | |||
BC066832.1 | - | 9 | 1243 | 30102312 |