Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002238.2 | NP_001002238.1 | 9 | 1140 | ||
NM_001002240.2 | NP_001002240.1 | 9 | 1113 | |||
NM_001002241.2 | NP_001002241.1 | 8 | 1413 | |||
NM_031387.3 | NP_113564.2 | 9 | 1273 | |||
GenBank mRNA | AB067571.1 | - | 8 | 1094 | ||
AB183526.1 | - | 9 | 1140 | |||
AB183527.1 | - | 9 | 1113 | |||
AB183528.1 | - | 8 | 1413 | |||
AB183529.1 | - | 8 | 977 | |||
AF285591.1 | - | 5 | 467 | |||
AK029806.1 | - | 9 | 1274 | |||
BC129954.1 | - | 4 | 399 | 40081321 | ||
BC129955.1 | - | 4 | 399 | 40081323 |