Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257096.1 | NP_001244025.1 | 3 | 1100 | ||
NM_006192.4 | NP_006183.2 | 3 | 1100 | |||
GenBank mRNA | AK303335.1 | - | 2 | 981 | ||
BC069134.1 | - | 2 | 764 | 7216990 | ||
BC143791.1 | - | 3 | 848 | |||
BC143793.1 | - | 3 | 845 |