Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031403.3 | NP_113580.2 | 7 | 1020 | ||
GenBank mRNA | AK050090.1 | - | 7 | 1021 | ||
AK090351.1 | - | 7 | 996 | |||
AK139631.1 | - | 7 | 1004 | |||
BC006661.1 | - | 7 | 1015 | 3501390 |