Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr137b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF154337.1 | - | 2, 3 | 507 | ||
AK009736.1 | - | 2, 3 | 509 | |||
AK010724.1 | - | 2, 3 | 522 | |||
AK170103.1 | - | 2, 3 | 517 | |||
AK170650.1 | - | 2, 3 | 425 | |||
AK171335.1 | - | 2, 3 | 517 | |||
BC093499.1 | - | 2, 3 | 504 |
Gene Symbol: Gpr137b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031999.2 | NP_114388.2 | 2, 3 | 544 | ||
GenBank mRNA | AK050369.1 | - | 2, 3 | 485 | ||
AK080884.1 | - | 2, 3 | 497 | |||
AK128965.1 | - | 2, 3 | 544 | |||
AK147921.1 | - | 2, 3 | 563 | |||
AK172286.1 | - | 2, 3 | 489 | |||
BC003212.1 | - | 2, 3 | 537 | 3157256 |