Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pard3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013580.3 | NP_001013598.1 | 8, 9 | 1296 | ||
NM_001013581.2 | NP_001013599.1 | 8, 9 | 1336 | |||
NM_001122850.1 | NP_001116322.1 | 10, 11 | 1845 | |||
NM_033620.2 | NP_296369.2 | 10, 11 | 1845 | |||
GenBank mRNA | AK037015.1 | - | 2, 3 | 177 | ||
AK052416.1 | - | 10, 11 | 1832 | |||
AK053563.1 | - | 10, 11 | 1826 | |||
AK053576.1 | - | 10, 11 | 1803 | |||
AK053762.1 | - | 10, 11 | 1831 | |||
AK137690.1 | - | 10, 11 | 1804 | |||
AY026057.1 | - | 10, 11 | 1656 | |||
AY856081.1 | - | 8, 9 | 1340 | |||
AY856082.1 | - | 8, 9 | 1336 | |||
BC090616.1 | - | 10, 11 | 1845 | 30943099 | ||
BC141187.1 | - | 10, 11 | 1607 | 9055799 | ||
BC145508.1 | - | 10, 11 | 1607 | 9054071 | ||
DQ082726.1 | - | 4, 5 | 713 |