Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adarb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289530.1 | NP_001276459.1 | 10 | 3195 | ||
NM_052977.5 | NP_443209.2 | 10 | 3200 | |||
GenBank mRNA | AK033916.1 | - | 10 | 3139 | ||
AK045538.1 | - | 10 | 3140 | |||
AK046083.1 | - | 10 | 3139 | |||
AK134964.1 | - | 10 | 3202 | |||
AK139079.1 | - | 10 | 3158 | |||
AK139273.1 | - | 10 | 3158 | |||
BC052426.1 | - | 10 | 3125 | 6835942 |