Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spock2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052994.2 | NP_443720.1 | 11 | 1763 | ||
GenBank mRNA | AK122233.1 | - | 11 | 1425 | ||
AK140483.1 | - | 11 | 1670 | |||
BC057324.1 | - | 11 | 1763 | 6821449 | ||
BC060210.1 | - | 9 | 1405 | 6838067 |