Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpnmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053110.4 | NP_444340.3 | 2 | 333 | ||
GenBank mRNA | AF322054.1 | - | 2 | 171 | ||
AJ251685.1 | - | 2 | 218 | |||
AK044764.1 | - | 2 | 227 | |||
AK076347.1 | - | 2 | 202 | |||
AK079220.1 | - | 175 | ||||
AK149700.1 | - | 2 | 202 | |||
AK149740.1 | - | 2 | 240 | |||
AK150739.1 | - | 2 | 202 | |||
AK154617.1 | - | 2 | 237 | |||
AK159165.1 | - | 2 | 232 | |||
AK159270.1 | - | 2 | 240 | |||
AK159747.1 | - | 2 | 202 | |||
AK159908.1 | - | 2 | 208 | |||
AK170506.1 | - | 2 | 232 | |||
AK171347.1 | - | 2 | 208 | |||
AK171441.1 | - | 2 | 202 | |||
AK171465.1 | - | 2 | 202 | |||
AK171474.1 | - | 2 | 202 | |||
AK171517.1 | - | 2 | 320 | |||
AK171608.1 | - | 2 | 202 | |||
AK171618.1 | - | 2 | 202 | |||
BC026375.1 | - | 2 | 207 | 4164706 | ||
BY205729.1 | - | 2 | 333 |